missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Total antibody (IgG and IgM) SARS-CoV-2 N protein ELISA Kit
Shop All Proteintech ProductsDescription
- KE30005 is a qualitative measurement of total antibody (IgG and IgM) to SARS-COV-2 N protein in serum or plasma.
- The principle of the kit is capture ELISA.
- Anti-human antibody has been pre- coated onto microplate well.
- The samples or standard are added to the well, after incubation the wells are washed and a horseradish peroxidase conjugated anti-N protein is added to each well.
- Producing an complex anti-human antibody and sample and SARS-CoV-2 N protein-HRP conjugated
- After incubation the wells are washed, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm
Specifications
Specifications
| Accession Number | NC_045512 |
| Assay Range | 2 to 128 ng/mL |
| Certifications/Compliance | ISO 13485 |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Immunoassay Kit Grade | Mix and Match Panel |
| Interassay CV | <10% |
| Intraassay CV | <10% |
| Shipping Condition | Blue ice |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Total antibody(IgG and IgM) SARS-CoV-2 N pr
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?