missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Total antibody (IgG and IgM) SARS-CoV-2 S-RBD ELISA Kit

Product Code. 16881656 Shop All Proteintech Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Type unitSize
16881656 Plasma, Serum 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16881656 Supplier Proteintech™ Supplier No. KE3000696T
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

KE30006 is a quantitative measurement of total antibody (IgG and IgM) to SARS-COV-2 S-RBD protein in serum or plasma

  • KE30006 is a quantitative measurement of total antibody (IgG and IgM) to SARS-COV-2 S-RBD protein in serum or plasma.
  • The principle of the kit is capture ELISA.
  • Anti-human antibody has been pre-coated onto microplate well.
  • The samples or standard are added to the well, after incubation the wells are washed and a horseradish peroxidase conjugated S-RBD protein is added to each well.
  • Producing an complex anti-human antibody and sample and SARS-CoV-2 S-RBD protein-HRP conjugated
  • After incubation the wells are washed, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.

Specifications

Accession Number NC_045512
Assay Range 6.25 to 200 ng/mL
Certifications/Compliance ISO 13485
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Immunoassay Kit Grade Mix and Match Panel
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
UniProt ID P0DTC2
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Total antibody (IgG and IgM) to SARS-CoV-
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.