Learn More
Hach Lange™ Total Nitrogen Reagent Set, LR, TNT
Description
For determination of Total Nitrogen by the Persulfate Digestion Test 'N Tube method: Hach Method 10071. Range: 0. 5 - 25. 0 mg/L N. 25-50 testsTest 'N Tube products provide safe, convenient testingEasy orderingPerform digestions and photometric measurements in the same vial.
Test 'N Tube products provide safe, convenient testing
Capped 16-mm vials provide a self-contained package for mixing and measurement.
Easy ordering
All necessary reagents and vials are contained in the package.
Perform digestions and photometric measurements in the same vial.
Save time, space and effort.
Specifications
Description: Nitrogen, total, TNT Reagent Set
Digestion Required: Yes
Footnote: †Dependent on the number of performed reagent blanks.
Method: 10071
Method Name: Persulfate Digestion
Number of tests: 25 - 50
Parameter: Nitrogen, total
Platform: Test 'N Tube™
Range: 0.5 - 25.0 mg/L N
Special Remarks: Set consists of 2 cartons: 26717-45 and 26721-45
26717-45 = TN Hydroxide reagent (2714045), Persulfate Powder Pillows (2671846)
26721-45 = TN Acid reagent C (2672145), Bisulfitreagent A Powder Pillows (2671946), Indicator reagent B Powder Pillows (2672046)
Storage conditions: 10 - 25 °C
Specifications
Specifications
| Type | Water Testing Kit Supplies and Reagents |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.