missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 92-600 Disposable Chemical Resistant Nitrile Gloves

Product Code. 11716584 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 100
Pack of 90
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11716584 Medium Pack of 100
16615135 X-Small Pack of 100
11706714 Small Pack of 100
11726584 Large Pack of 100
11786584 X-Large Pack of 100
32078683 2X-Large Pack of 90
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11716584 Supplier Ansell Supplier No. 92600080
  • ONLINE EXCLUSIVE PRICE
    294.80 €
    18.46 € / Pack of 100
    Save 276.34 €
    93% Off
View all promotions
In Stock
Add to basket
Edge
Ajouter au panier

missing translation for 'validMainframeMsg'

Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.

  • Made from 100% nitrile
  • Resistant to nuisance punctures, nicks, cuts and abrasions
  • Comfortable thin mil construction
  • Rolled cuffs for easy donning and doffing
  • Chemical resistant
TRUSTED_SUSTAINABILITY

Spécification

Certifications/Compliance ASTM D6319, FDA 21 CFR 177.2600, ASTM F1671
Certifications/Compliance (Europe) Category III, EN 420:2003 + A1:2009
Chlorinated No
Applications Industrial
Color Green
Disposable Single-use
Finger Thickness (English) 5.5 mil
Finger Thickness (Metric) 0.14 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.7 mil
Palm Thickness (Metric) 0.12 mm
Product Type Chemical-resistant Gloves
Size Medium
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
EN 421 No
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKPT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Beaded/Rolled
Lining Unlined
Manufacturing/QMS Audit Standards ISO 9001
Model 92 600 M 585835
Series TouchNTuff™ 92-600
Length (English) 9.5 in.
Length (Metric) 24 cm
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.