missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 92-500 PPE Gloves

Product Code. 17747154
Change view
Click to view available options
Size:
7
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17747154 7 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17747154 Supplier Ansell Supplier No. 92500070

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Powdered gloves offering proven protection against hazardous chemical splashes.

  • Superior protection: Made with TNT™ Chemical Splash Resistance Technology, these gloves effectively safeguard hands from exposure to harmful chemicals
  • Improved comfort: Soft nitrile materials enhances comfort during extended wear
  • Robust durability: Strong proprietary formulation prevents tearing during heavy-duty works
  • Powdered design: TouchNTuff™ 92-500 PPE gloves are powdered for moisture absorption and easy donning
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Color Green
Finger Thickness (English) 5.5 mil
Finger Thickness (Metric) 0.14 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.3 mil
Palm Thickness (Metric) 0.11 mm
Product Type Chemical-resistant Gloves
Size 7
Thickness (English) 4.3 mil
Thickness (Metric) 0.11 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.