missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 93-163 Series Blue Disposable Nitrile Gloves

Product Code. 15522505
Click to view available options
Size:
10.5/11
6.5/7
7.5/8
8.5/9
9.5/10
Unit Size:
Pack of 50
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 15522505

Brand: Ansell 93163100_cleaned

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Proven splash resistance against hazardous chemicals. Ansell TouchNTuff™ 93-163 Series Blue Disposable Nitrile Gloves are flexible and comfortable offering better dexterity.

  • Single use gloves
  • Nitrile formulation for low hand fatigue
  • Easy donning
  • Textured finish, light and resistant
  • Good resistance to puncture
  • Flexible and comfortable offering excellent dexterity
  • Beaded cuff for an easy donning and offering good tear resistance
  • Ambidextrous

Applications:

  • Chemical handling
  • Glass manufacturing
  • Handling of cytostatics
  • Intricate parts handling
  • Laboratory analysis
  • Light assembly of oil-coated pieces
  • Light assembly tasks
  • Paint and spray shops
  • Pharmaceuticals
  • Printing industry
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 374:2003
Accelerator Free Yes
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size 9.5/10
Sterility Non-sterile
Thickness (Metric) 0.17 mm
Glove Cuff Style Beaded/Rolled
Packaging Type 50 gloves in a box, 10 boxes in a carton
Series 93-163
Length (Metric) 355 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.