missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ TouchNTuff™ 93-300 Series Green Nitrile Gloves
BIOPHARMA

Product Code. 15254698
Change view
Click to view available options
Size:
10
9.5
Large
Medium
Small
X-Large
Unit Size:
Pack of 100
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15254698 Medium Pack of 1000
15294698 10 Pack of 1000
15284698 X-Large Pack of 1000
15201588 9.5 Pack of 100
15274698 Large Pack of 1000
15366195 Small Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15254698 Supplier Ansell™ Supplier No. 93300080
  • ONLINE EXCLUSIVE PRICE
    675.40 €
    436.92 € / Pack of 1000
    Save 238.48 €
    35% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

Complies with ASTM D 3577, Type II. Ansell™ TouchNTuff™ 93-300 Series Green Nitrile Gloves are ideal for use in electronics and semiconductor manufacturing, and laboratory testing.

  • Delivers the highest protection for a wide range of chemicals than any other single use nitrile glove
  • Provides the wearer with a high level of comfort and increased protection in the forearm (longer cuff)
  • Manufactured from nitrile, offering three times the puncture resistance of standard latex gloves
  • Made without natural rubber latex proteins, the glove may be worn with confidence by latex sensitive (Type l) wearers
  • Suitable for ISO Class 5 or higher clean room environments
TRUSTED_SUSTAINABILITY

Specifications

Back Material Nitrile
Certifications/Compliance Standards CATEGORY III EN 374, EN 421, ASTM D 3577, Type II
Cleanroom Class Class 100 (ISO 5)
Cuff Material Nitrile
Accelerator Free No
AQL 1.5
Coating Material Nitrile
Color Green
Disposable Single-use
Exterior Finish Fingertip Textured
Hand Specific
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Controlled Environment Gloves
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.12 mm
Finger Style Textured fingertips
For Use With (Application) For blending of liquids, filtration processes, vaccines injectables operations etc.
Quantity 240 Pairs
Resistant To Splash and high puncture resistance
Series 93-300
Show More Show Less
Product Title
Select an issue

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.