missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ TouchNTuff™ Extended Cuff Disposable Green Nitrile Gloves

Product Code. 11746733
Change view
Click to view available options
Size:
6.5, 7
7.5, 8
8.5, 9
9.5, 10
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11746733 6.5, 7 Pack of 100
11756733 7.5, 8 Pack of 100
12465110 8.5, 9 Pack of 100
11776733 9.5, 10 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11746733 Supplier Ansell™ Supplier No. 92605070
Spotlight Offers
Use promo code 28036
View all promotions
In Stock
Add to basket
Edge
Add to basket

Offers up to four times the puncture resistance of comparable natural-latex gloves, and three times the resistance of similar neoprene gloves. Resists a greater variety of industrial chemicals for longer periods than any other nitrile disposable gloves.

  • Contain no natural rubber proteins, which means no risk of Type I allergies for the wearer.
  • Easy donning and strong grip in wet or dry conditions with a unique Thin Nitrile Technology formulation
  • Highly versatile and suitable for many different uses.
TRUSTED_SUSTAINABILITY

Specifications

Classification PPE Category III
Accelerator Free No
Applications Industrial
AQL 1.5
Color Green
Disposable Single-use
Exterior Finish Fingertip Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Touch N Tuff Gloves
Size 6.5, 7
Sterility Non-sterile
Style Beaded Cuff
Thickness (Metric) 0.12 mm
Glove Cuff Style Beaded/Rolled
Quantity 100 Pack
Length (Metric) 300 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.