missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ TouchNTuff™ Extended Cuff Disposable Green Nitrile Gloves

Product Code. 11776733 Shop All Ansell Products
Change view
Click to view available options
Size:
6.5, 7
7.5, 8
8.5, 9
9.5, 10
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11776733 9.5, 10 Pack of 100
11746733 6.5, 7 Pack of 100
11756733 7.5, 8 Pack of 100
12465110 8.5, 9 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11776733 Supplier Ansell™ Supplier No. 92605100
  • ONLINE EXCLUSIVE PRICE
    36.60 €
    22.89 € / Pack of 100
    Save 13.71 €
    37% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Offers up to four times the puncture resistance of comparable natural-latex gloves, and three times the resistance of similar neoprene gloves. Resists a greater variety of industrial chemicals for longer periods than any other nitrile disposable gloves.

  • Contain no natural rubber proteins, which means no risk of Type I allergies for the wearer.
  • Easy donning and strong grip in wet or dry conditions with a unique Thin Nitrile Technology formulation
  • Highly versatile and suitable for many different uses.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Category III, EN374, EN 420, EN374/1, 2, 3, ASTM D 6319, FDA 21 CFR 177-2600, Areete 09/11/04, European Regulation 1935/2004, German Recommendation for Food Commodities
Classification PPE Category III
Accelerator Free No
Applications Industrial
AQL 1.5
Coating Material Polymer
Color Green
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 0.0063 in.
Finger Thickness (Metric) 0.16 mm
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Nitrile
Palm Thickness (English) 0.12 mm
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Product Type Gloves
Size 9.5, 10
Sterility Non-sterile
Style Rolled beaded
Thickness (English) 0.0063 in.
Thickness (Metric) 0.16 mm
Finger Style Textured
For Use With (Application) Ideal applications chemical handling, glass manufacturing, intricate parts handling, light assembly of oil-coated pieces, paint and spray shops, printing industry, electronics, handling of cytostatics, laboratory analysis, light assembly tasks, pharmaceuticals
Inspected for Medical Use Yes
Quantity 100 Pk.
Resistant To Splash resistance against hazardous chemicals, puncture
Series 92-605
Tensile Strength 14 MPa
Length (Metric) 300 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.