Learn More
DiversifiedBiotech Tough-Spots™
GREENER_CHOICE
Product Code. 30282266
Change view
Click to view available options
Quantity:
1500/Pk.
500/Pk.
Unit Size:
Pack of 1500
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30282266 500/Pk. Pack of 500
30282276 1500/Pk. Pack of 1500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30282266 Supplier DiversifiedBiotech Supplier No. TSPOTS75
Only null left
Add to basket
Edge
Add to basket
Pre-cut peel off labels for tube tops with PVC cryogenic labels
  • Circular cryogenic labels formatted for handwriting
  • Tough Spots™ on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers
  • Tough Spots™ come in a variety of colors and sizes to ensure the right label for the right job!
  • Adheres to most plastics (100% virgin PP tubes recommended), glass and metals without cracking, peeling or degrading
  • Flexible polyolefin material and acrylic adhesive expand and contract with temperature fluctuations
  • Roll of 1007
Specifications
Adhesive Type Peel and Stick, Acrylic
Color White
Finish Semi-gloss
Format Roll
Material Polyolefin
Print Technology Laser
Product Type Ultra Reflective Letter Label
Sterility Non-sterile
Quantity 500/Pk.
Length (English) 0.75 in.
Width (English) 0.75 in.
Width (Metric) 1.9 cm