Learn More
Brady™ ToughBond™ B-483 Gloss Polyester Labels with an Aggressive Adhesive
For use in commercial/industrial applications. Brady B-483 gloss polyester labels with an aggressive adhesive is used for BMP71 printer.
Brand:  Brady™ 114807
Code : NEW
Additional Details : Weight : 0.08890kg
 View more versions of this product
Product Code. 11767274
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
UL Recognized, CSA Approved
Rubber, Permanent
Matte
Asset Tracking Label
BMP™71 Printer
250/Roll
1.5 x 0.75 in. (W x H)
-40°C to +120°C
0.102 mm
19.05 x 38.10 mm
19.05 mm
Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant
White
Polyester (B-483)
Component ID, Equipment ID, Barcoding, Panel ID, Voice/Data ID, Asset tracking, Rating plates, General purpose ID
1
Rectangle
38.1 x 19.1 mm (W x H)
0.004 in.
22.23 mm
38.10 mm