missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ToxBLAzer™ DualScreen Kit

Product Code. 10624223
Change view
Click to view available options
Quantity:
200 μg
5mg
Unit Size:
5mg
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10624223 200 μg Each
11504936 5mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10624223 Supplier Invitrogen™ Supplier No. K1138
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ToxBLAzer™ DualScreen Kit

ToxBLAzer™ DualScreen Kit provide the key advantage of combining the ratiometric reporter gene readout of beta-lactamase with a cytotoxicity readout in the same assay. Thus, ToxBLAzer™ DualScreen yields reporter readouts that normalize cytotoxic cellular responses, which is a necessity in screening applications which down regulate transcription. The ToxBLAzer™ substrate contains both the LiveBLAzer™-FRET B/G substrate and a proprietary cytotoxicity indicator. Both compounds readily enter the cell, where endogenous esterases rapidly convert them into fluorescent compounds Esterase activity and an intact cell membrane are required for detection of the indicator. The cytotoxicity indicator's fluorescence is red-shifted to ensure compatibility with the blue and green emissions of the LiveBLAzer™-FRET B/G substrate, allowing detection of both reporter gene activity and cytotoxicity in the same assay sample.

Advantages of ToxBLAzer™ DualScreen Kits:

  • Multiplexed readout results in rapid identification of false positives due to toxicity or lack of viable cells, thereby improving hit-picking efficiency
  • Ratiometric blue:green reporter readout reduces false hits by minimizing experimental noise
  • Single reagent addition eliminates need for follow up cytotoxicity assays, thereby increasing throughput while reducing sample and test compound consumption
  • TRUSTED_SUSTAINABILITY

    Specifications

    For Use With (Equipment) Microplate Reader, Fluorometer, Flow Cytometer
    Product Line ToxBLAzer
    Content And Storage ToxBLAzer™ DualScreen Loading Kits include ToxBLAzer™ substrate, DMSO (for Solution A), Solution B, and Solution C. ToxBLAzer™ substrate is provided lyophilized and should be stored at 20°C, dessicated and protected from light. DMSO (for Solution A), Solution B, and Solution C should be stored at 22-25°C, protected from direct light.

    The 200 μg kit is sufficient for 3,750 x 48 μl standard protocol well assays; the 5 mg kit is sufficient for 93,750 x 48 μl standard protocol well assays. Bulk quantities of loading kits are available upon request.
    Detection Method Fluorescence
    Product Type Dual Screen Kit
    Quantity 200 μg
    Format Kit
    Cell Permeability Cell-Permeant
    Substrate Beta-Lactamase Substrate

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.