missing translation for 'onlineSavingsMsg'
Learn More

Toyocamycin, TRC

Product Code. 30452933
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452933 5 mg 5mg
30450375 25 mg 25mg
30452679 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452933 Supplier Toronto Research Chemicals Supplier No. TRCT7000035MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-58-6
Molecular Formula C12H13N5O4
Molecular Weight (g/mol) 291.26
Synonym 4-Amino-7-beta-D-ribofuranosyl-7H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile, 4-Amino-5-cyano-7-(beta-D-ribofuranosyl)pyrrolo[2,3-d]pyrimidine, 7-Cyano-7-deazaadenosine, Antibiotic 1037, Antibiotic E 212, MT 1155, NSC 63701NSC 99843, SIPI 763-1, Toyocamycin Nucleoside, Unamycin B, Vengicide
IUPAC Name 4-amino-7-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrrolo[2,3-d]pyrimidine-5-carbonitrile
SMILES Nc1ncnc2c1c(cn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C#N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H13N5O4/c13-1-5-2-17(11-7(5)10(14)15-4-16-11)12-9(20)8(19)6(3-18)21-12/h2,4,6,8-9,12,18-20H,3H2,(H2,14,15,16)/t6-,8-,9-,12-/m1/s1
Formula Weight 291.0968
Chemical Name or Material Toyocamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.