missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ TR-FRET Progesterone Receptor Coactivator Assay Kit

Product Code. 15657858
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15657858 800 x 20 μL assays Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15657858 Supplier Invitrogen™ Supplier No. A15903
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LanthaScreen™ TR-FRET Progesterone Receptor Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential progesterone receptor (PR) ligands as agonists or antagonists of ligand-dependent coactivator recruitment.

  • The LanthaScreen™ TR-FRET Progesterone Receptor Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential progesterone receptor (PR) ligands as agonists or antagonists of ligand-dependent coactivator recruitment. The kit uses a PR ligand--binding domain (PR-LBD) tagged with glutathione-S-transferase (GST) (also available separately), a terbium (Tb)-labeled anti-GST antibody, and a fluorescein-labeled coactivator peptide in a homogenous mix-and-read assay format.
  • Agonist Mode - When using the LanthaScreen TR-FRET Progesterone Receptor Coactivator Assay in agonist mode (to identify agonist compounds), PR-LBD is added to ligand test compounds, followed by the addition of a mixture of fluorescein-labeled coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the TR-FRET ratio of 520/495 nm is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the PR-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide.
  • Antagonist Mode - When using the LanthaScreen™ TR-FRET Progesterone Receptor Coactivator Assay in antagonist mode (to identify antagonist compounds), PR-LBD is added to ligand test compounds followed by addition of a mixture of agonist, fluorescein-labeled coactivator peptide, and terbium-labeled anti-GST antibody. The concentration of agonist used in this mode is the EC80 concentration as determined by first running the assay in agonist mode.
TRUSTED_SUSTAINABILITY

Tekniske data

Content And Storage 1 tube PR Ligand Binding Domain Recombinant Protein (store at -68 to -85°C)
1 tube of Peptide (store at -5 to -30°C)
2 bottles TR-FRET Buffer (store at -5 to -30°C)
1 tube Tb-Anti-GST Ab (store at -5 to -30°C)
1 tube of DTT (store at -5 to -30°C)
For Use With (Application) Co-Factor Interaction Assay, Nuclear Receptor Assay
Product Line LanthaScreen
Quantity 800 x 20 μL assays
Type Coactivator Assay Kit

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.