missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Digital Dial Thermometer

Easy to read digital dial and mercury free. Fisherbrand Traceable Digital Dial Thermometers are ideal for cuvets, soil, incubators and test tubes. Wide temperature span combines the ranges of more than 90 different mercury thermometers.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.11000kg

 View more versions of this product

Product Code. 15223986

  • 46.30 € / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Includes: Protective case, Silver Oxide battery

Description

Description

  • LCD display 20 times more readable than eye-straining pointers or mercury levels
  • Readings updated every 10 seconds
  • Operate continuously for 1.5 years on a single replaceable battery
  • Resolution, accuracy: 0.10, ±1°C Stem length: 4.8 in. (12 cm)
  • Stem diameter: 0.12 in. (3mm)
  • Dial diameter: 1.5 in. (38mm)
  • Weight: 1.01 oz. (31g)
  • Range: -58 to 572°F and -50 to 300°C

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Specifications

ISO 17025, A2LA, NIST
Soil, cuvettes, test tubes, food products, incubators, biomedical reagents and petroleum Hydrocarbons
4.8 in.
Plastic
1 oz.
Thermometer with centigrade and fahrenheit scale
300°C
-50°C
-50°C to +300°C
Digital LCD
0.19 in.
Protective case, Silver Oxide battery
12 cm
0.1°
31 g
572°F
-58°F
-58°F to +572°F
±1°C/±1°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers