missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ TraceableGO™ Temperature Data Logger with Bluetooth™, Calibration and Bottle Probe
Shop All Fisherbrand Traceable ProductsDescription
- Critical Information (Temperature, Trip Duration, MIN/MAX, Battery Level and Alarms) displayed on LCD at all times Simple, single button use
- Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location
- Compatible with TraceableGO or TraceableLIVE Mobile Apps:
- Once the trip has ended, users may export the data
- App immediately identifies if the contents experienced conditions outside user-defined parameters
- View if an alarm was triggered
- Generate an instant data report
- Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours
- Logging duration is 227 days (7.5 months)
- TraceableGO™ Bluetooth devices are pre-programmed with alarm values
Specifications
Specifications
| Type | Temperature Datalogger |
| Display Type | Multi-line LCD |
| Data Memory | 64,000 Readings, 7.5 Months, 5 min. Interval |
| Alarm Types | High/Low |
| Software Type | TraceableLIVE |
| Battery Type | 2 AAA |
| Description | Bluetooth datalogging thermometer with bottle probe |
| Includes | Batteries, wall mount, Traceable™ Certificate |
| Channels | 1 |
| Certifications/Compliance | ISO 17025, NIST |
| Show More |
Option to subscribe to cloud-based TraceableLIVE™ service to upload data to cloud.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?