Learn More
3M™ Protective Coverall 4520
Product Code. 15710068 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15710068 Medium Pack of 20
15720068 Large Pack of 20
15730068 2X-Large Pack of 20
15700068 X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15710068 Supplier 3M™ Supplier No. 7100048967
  • ONLINE EXCLUSIVE PRICE
    198.00 €
    124.01 € / Pack of 20
    Save 73.99 €
    37% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Coverall has been engineered to offer multiple layers of SMMMS for effective hold out and strength at a lightweight 43 gsm. Breathable and soft with a cloth like feel, it can be worn for extended periods while providing protection against static discharge, hazardous particulates, and liquid splash Type 5/6.
  • Type 5/6 lightweight and breathable coverall, offering protection against hazardous dusts and limited liquid splashes
  • Standard collar provides comfort and convenience when a hood is not needed
  • Knitted cuffs with elastic waist and ankles for convenience and freedom of movement
  • Two-way zipper with storm flap for convenience and to help provide additional protection
  • Anti-static coating on both sides helps reduce static build-up
  • Robust, low-linting, highly breathable 43 gsm SMMMS material
  • Meets inward leakage requirements of EN 1073-2 for protection against nuclear particles
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Medium
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Cuff Style Knit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance CE Category III, EN 1073-2 Particulate Radioactive Contamination, EN 1149-5 - Electrostatic Properties, EN 13034:2005+A1:2009 Type 6 Liquid Splash, EN ISO 13982-1:2004 Type 5 Hazardous Dusts
Certifications/Compliance (Europe) CE Category III, EN 1073-2, EN 1149-5, EN 13034:2005+A1:2009 Type 6, EN ISO 13982-1:2004 Type 5