missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ TraceableOne™ 10 Pack Single-User Datalogger

Product Code. 16089511 Shop All Fisherbrand Products
Change view
Click to view available options
Description:
Preprogrammed -20°C to -15°C
Preprogrammed 2°C to 8°C
Alarm Types:
Preprogrammed -20°C to -15°C
Preprogrammed 2°C to 8°C
Data Memory:
16,000 Readings, 10 Day, 1 min. Interval
16,000 Readings, 30 Day, 3 min. Interval
16,000 Readings, 60 Day, 6 min. Interval
16,000 Readings, 90 Day, 10 min. Interval
Unit Size:
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Alarm Types Data Memory unitSize
16089511 Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C 16,000 Readings, 10 Day, 1 min. Interval Pack of 10
16059521 Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C 16,000 Readings, 90 Day, 10 min. Interval Pack of 10
16039521 Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C 16,000 Readings, 60 Day, 6 min. Interval Pack of 10
16049521 Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C 16,000 Readings, 90 Day, 10 min. Interval Pack of 10
16009521 Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C 16,000 Readings, 30 Day, 3 min. Interval Pack of 10
16099511 Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C 16,000 Readings, 10 Day, 1 min. Interval Pack of 10
16019521 Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C 16,000 Readings, 30 Day, 3 min. Interval Pack of 10
16029521 Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C 16,000 Readings, 60 Day, 6 min. Interval Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 16089511

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use logger right out of the box to track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals and food items during transit. Fisherbrand™ TraceableOne™ 10 Pack Single-User Datalogger allows for monitoring materials every step of the way.

  • Logger generates PDF report
  • Download directly to PC
  • No Software required
  • Compact
  • Single-use
  • Flashing Red and Green LED's indicate if logger is recording data, has stopped recording and if an alarm has been triggered
  • Press start to begin recording crucial temperature data, 30 minute time delay before logging begins

Specifications

Type Datalogger
Data Memory 16,000 Readings, 10 Day, 1 min. Interval
Calibration Calibrated
Alarm Types Preprogrammed 2°C to 8°C
Software Type No software required, download data as PDF report
Description Preprogrammed 2°C to 8°C
Includes Batch-calibrated Traceable certificate
IP Rating IP67
For Use With (Application) Refrigerated transportation samples
Accuracy (Temperature) ±0.5°C from -20° to 40°C, otherwise ±1°C
Temperature Resolution 0.01°
Temperature Range (Metric) -30°C to +70°C
Length (Metric) 8.89 cm
Electrical Requirements Battery-Powered
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.