Learn More
Fisherbrand™ TraceableOne™ Single-Use USB Temperature Data Logger
The TraceableOne™ Single-Use USB Temperature Data Logger automatically generates PDF data report via USB port.
Brand:  Fisherbrand™
Additional Details : Weight : 0.91170kg
 View more versions of this product
Product Code. 17357619
  • 1090.00 € / Pack of 40
Estimated Shipment: 16-07-2024
to see stock.
Description
Description
  • LED alarm indicator for visual status indication
  • Time delay of 30 minutes before logging begins for enhanced logging accuracy
  • Protective plastic pouch maintains waterproof IP67 rating
  • Bar code for in-house scanning
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Specifications
16,000 Readings, 30 Day, 3 Minute Interval
2°C to 8°C