missing translation for 'onlineSavingsMsg'
Learn More

Tragacanth powder

Product Code. 11458417
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458417 100 g 100g
11468417 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458417 Supplier Thermo Scientific Alfa Aesar Supplier No. A18502.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 9000-65-1

Tragacanth powder is used as an emulsifier, thickener, stabilizer in food and pharmaceutical industry. It acts as a binder used in the preparation of artist's pastels. It is used as a natural hair shampoo. It plays an important role as freezing-embedding medium for tissue preservation. It is used as a demulcent in cough and cold preparations as well as to manage diarrhea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-65-1
MDL Number MFCD00131255
Synonym Gum tragacanth

Specifications

Merck Index 14,4581
Solubility Information Slightly soluble in water.
Quantity 100 g
Odor Odorless
Chemical Name or Material Tragacanth powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.