missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRAM 34

Product Code. 15432869
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432869 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15432869 Supplier Thermo Scientific Chemicals Supplier No. J60019.MB
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 289905-88-0 | C22H17ClN2, C22H17ClN2 | 344.84 g/mol

TRAM-34 blocks intermediate conductance calcium-activated potassium channel IKCa1 with a Kd of 20nM and exhibits exquisite selectivity for the channel. Clotrimazole impurity. Also used to study its effect on the prevention of vascular restenosis after balloon angioplasty.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Description Yellow
Content And Storage 2°C to 8°C
For Use With (Application) TRAM-34 is a potent inhibitor of the intermediate-conductance calcium-activated potassium channel
CAS 289905-88-0
Molecular Formula C22H17ClN2
Formula Weight 344.84g/mol
Synonym 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole
Melting Point 145°C to 147°C
Solubility Soluble in DMSO at 2mg/ml; Insoluble in water
Product Type Signal Transduction Reagent
Form Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.