missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TRAM 34

Product Code. 30002117
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002117 10 mg 10mg
18084082 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002117 Supplier R&D Systems™ Supplier No. 2946/10
  • ONLINE EXCLUSIVE PRICE
    245.54 €
    214.20 € / 10mg
    Save 31.34 €
    12% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and highly selective KCa3.1 channel blocker

Potent and highly selective blocker of intermediate conductance Ca2+-activated K+ channels (KCa3.1) (Kd = 20 nM). Exhibits 200-1500-fold selectivity over KV, BKCa, KCa2, Na+, CRAC and Cl- channels. Suppresses the reactivation of lymphocytes by mitogenic stimuli.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 289905-88-0
Molecular Formula C22H17ClN2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 344.84
Purity >99%
Molecular Weight (g/mol) 344.842
Chemical Name or Material TRAM 34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.