missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Trametinib
GREENER_CHOICE

Product Code. 18755836
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18755836 50 mg 50mg
18746376 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18755836 Supplier Tocris Bioscience™ Supplier No. 7709/50
  • ONLINE EXCLUSIVE PRICE
    582.00 €
    550.20 € / 50mg
    Save 31.80 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Potent and selective MEK inhibitor; orally bioavailable

Trametinib is a potent and selective MEK1 and MEK2 inhibitor (IC50 = 0.92 and 1.8 nM, respectively, in cell-free assays). In vitro, it inhibits the growth of human colorectal cancer cell lines expressing either B-RAF or K-RAF (IC50 values range from 0.48 to 36 nM). In vivo, it suppresses tumor growth of HT-29 and COLO205 xenografts in nude mice. Trametinib induces apoptosis. This compound is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-[3-Cyclopropyl-5-[(2-fluoro-4-iodophenyl)amino]-3,4,6,7-tetrahydro-6,8-dimethyl-2,4,7-trioxopyrido[4,3-d]pyrimidin-1(2 H)-yl]phenyl]acetamide
CAS 871700-17-3
Quantity 50 mg
Target MEK Inhibitors
Molecular Formula C26H23FIN5O4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.