missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tranilast, ACROS Organics™

Brand:  Thermo Scientific™ 468610050

Additional Details : CAS Number : 53902-12-8 Weight : 0.00500kg

 View more versions of this product

Product Code. 17302491

  • 206.00 € / 5g
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Tranilast
5 g
9646
C18 H17 N O5
Crystalline Powder
53902-12-8
insoluble in water
166.2°C to 168.2°C
327.33
White to Light-yellow to Green
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers