missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Bis(diphenylphosphino)ethylene, 98%

Product Code. 11328205
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328205 5 g 5g
11318205 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328205 Supplier Thermo Scientific Alfa Aesar Supplier No. L08508.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 983-81-3 | C26H22P2 | 396.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 983-81-3
Molecular Formula C26H22P2
Molecular Weight (g/mol) 396.41
MDL Number MFCD00003046 MFCD00063219
InChI Key NCKJIJSEWKIXAT-QURGRASLSA-N
Synonym trans-1,2-bis diphenylphosphino ethylene, e-1,2-bis diphenylphosphino ethene, ethene-1,2-diylbis diphenylphosphane, e-vinylenebis diphenylphosphine, cis-1,2-bis diphenylphosphino ethylene, trans-1,2-ethenediylbis diphenylphosphine, trans-vinylenebis diphenylphosphine, cis-vinylenebis diphenylphosphine
PubChem CID 2733288
IUPAC Name [(E)-2-diphenylphosphanylethenyl]-diphenylphosphane
SMILES C(=C/P(C1=CC=CC=C1)C1=CC=CC=C1)\P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 126°C to 130°C
Quantity 5 g
Beilstein 757793
Sensitivity Air Sensitive
Formula Weight 396.41
Percent Purity 98%
Chemical Name or Material trans-1,2-Bis(diphenylphosphino)ethylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.