missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Bis(tri-n-butylstannyl)ethylene, 95%

Product Code. 10310584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10310584 1 g Glass bottle 1g
10249053 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10310584 Proveedor Thermo Scientific Acros N.º de proveedor 388670010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 14275-61-7 | C26H56Sn2 | 606.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 14275-61-7
Molecular Formula C26H56Sn2
Molecular Weight (g/mol) 606.15
MDL Number MFCD01631299
InChI Key VNKOWRBFAJTPLS-UHFFFAOYSA-N
Synonym trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, e-1,2-bis-tri-n-butylstannyl-ethylene, tributyl 2-tributylstannyl ethenyl stannane
PubChem CID 5378370
IUPAC Name tributyl-[(E)-2-tributylstannylethenyl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC

Especificaciones

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3(CH2)3)3SnCHCHSn((CH2)3CH3)3
Refractive Index 1.5000 to 1.5030 (20°C, 589nm)
Quantity 1 g
Formula Weight 606.15
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material trans-1, 2-Bis(tri-n-butylstannyl)ethylene
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • trans-1,2-Bis(tri-n-butylstannyl)ethylene
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Indicaciones de peligro
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Consejos de prudencia
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.