missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Bis(tri-n-butylstannyl)ethylene, 96%

Product Code. 11374576
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374576 1 g 1g
11384576 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374576 Supplier Thermo Scientific Alfa Aesar Supplier No. L20130.03
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14275-61-7 | C26H56Sn2 | 606.154 g/mol

trans-1,2-Bis(tri-n-butylstannyl)ethylene is used as an intermediate in organic synthesis. It is also employed in the synthesis of gamma, delta-alkynones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Bis(tri-n-butylstannyl)ethylene is used as an intermediate in organic synthesis. It is also employed in the synthesis of gamma, delta-alkynones.

Solubility
Miscible with terahydrofuran and dimethyl formamide. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14275-61-7
Molecular Formula C26H56Sn2
Molecular Weight (g/mol) 606.154
MDL Number MFCD01631299
InChI Key VNKOWRBFAJTPLS-UHFFFAOYSA-N
Synonym trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, e-1,2-bis-tri-n-butylstannyl-ethylene, tributyl 2-tributylstannyl ethenyl stannane
PubChem CID 5378370
IUPAC Name tributyl-[(E)-2-tributylstannylethenyl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC

Specifications

Density 1.14
Boiling Point 185°C (0.01 mmHg)
Refractive Index 1.502
Quantity 1 g
UN Number UN2788
Beilstein 3613803
Sensitivity Air and moisture sensitive
Solubility Information Miscible with terahydrofuran and dimethyl formamide. Slightly miscible with water.
Formula Weight 606.15
Percent Purity 96%
Chemical Name or Material trans-1,2-Bis(tri-n-butylstannyl)ethylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • trans-1,2-Bis(tri-n-butylstannyl)ethylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: trans-1,2-Bis(tributylstannyl)ethene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.