missing translation for 'onlineSavingsMsg'
Learn More

(±)-trans-1,2-Diaminocyclohexane, 98%

Product Code. 10038581 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10038581 50 mL Glass bottle 50mL
10170452 10 mL Glass bottle 10mL
10405282 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10038581 Supplier Thermo Scientific Acros Supplier No. 248670500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1121-22-8 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-22-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00063747
InChI Key SSJXIUAHEKJCMH-PHDIDXHHSA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 14.0°C to 15.0°C
Color Colorless to Yellow
Density 0.9510g/mL
Boiling Point 79.0°C to 81.0°C (15.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(NH2)2
Refractive Index 1.4885 to 1.4905
Quantity 50 mL
Specific Gravity 0.951
Solubility Information Solubility in water: soluble.
Formula Weight 114.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (±)-trans-1, 2-Diaminocyclohexane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (#1)-trans-1,2-Diaminocyclohexane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.