missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dibromocyclohexane, 99%, Thermo Scientific™

Product Code. 10715181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10715181 25 mL Glass bottle 25mL
10591554 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10715181 Supplier Thermo Scientific Acros Supplier No. 112770250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7429-37-0
Molecular Formula C6H10Br2
Molecular Weight (g/mol) 241.95
MDL Number MFCD00003820
InChI Key CZNHKZKWKJNOTE-PHDIDXHHSA-N
Synonym trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane
PubChem CID 98506
IUPAC Name (1R,2R)-1,2-dibromocyclohexane
SMILES Br[C@@H]1CCCC[C@H]1Br

Specifications

Color Yellow
Density 1.7800g/mL
Boiling Point 145°C (100.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H10Br2
Refractive Index 1.5510 to 1.553
Quantity 25 mL
Beilstein 05, 24
Specific Gravity 1.78
Formula Weight 241.95
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material trans-1, 2-Dibromocyclohexane, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.