missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dibromocyclohexane, 99%, Thermo Scientific Chemicals

Product Code. 15460288
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460288 100 g 100g
15440288 5 g 5g
15450288 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460288 Supplier Thermo Scientific Alfa Aesar Supplier No. H53456.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

trans-1,2-Dibromocyclohexane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Dibromocyclohexane is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-37-0
Molecular Formula C6H10Br2
Molecular Weight (g/mol) 241.95
MDL Number MFCD00003820
InChI Key CZNHKZKWKJNOTE-PHDIDXHHSA-N
Synonym trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane
PubChem CID 98506
IUPAC Name (1R,2R)-1,2-dibromocyclohexane
SMILES Br[C@@H]1CCCC[C@H]1Br

Specifications

Density 1.784
Boiling Point 145°C (100 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.5515
Quantity 100 g
Formula Weight 241.95
Percent Purity 99%
Chemical Name or Material trans-1,2-Dibromocyclohexane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.