missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichlorocyclohexane, TRC

Product Code. 30456686
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456686 100 mg 100mg
30449690 50 mg -
30448951 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456686 Supplier Toronto Research Chemicals Supplier No. TRCT705130100MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-86-6
Molecular Formula C6H10Cl2
Molecular Weight (g/mol) 153.05
IUPAC Name (1R,2R)-1,2-dichlorocyclohexane
SMILES Cl[C@@H]1CCCC[C@H]1Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H10Cl2/c7-5-3-1-2-4-6(5)8/h5-6H,1-4H2/t5-,6-/m1/s1
Formula Weight 152.016
Chemical Name or Material trans-1,2-Dichlorocyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.