missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichloroethylene, 98%, stab. with 4-methoxyphenol

Product Code. 11398865
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398865 25 g 25g
11308875 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398865 Supplier Thermo Scientific Alfa Aesar Supplier No. L13994.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 156-60-5 | C2H2Cl2 | 96.94 g/mol

trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.

Solubility
Miscible with ethanol, ethyl ether, acetone, benzene and chloroform. Immiscible with water.

Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-60-5
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.94
MDL Number MFCD00062942
InChI Key KFUSEUYYWQURPO-OWOJBTEDSA-N
Synonym trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
PubChem CID 638186
ChEBI CHEBI:29027
IUPAC Name (E)-1,2-dichloroethene
SMILES Cl\C=C\Cl

Specifications

Melting Point -50°C
Density 1.252
Boiling Point 47°C to 49°C
Flash Point 2°C (35°F)
Refractive Index 1.4455
Quantity 25 g
UN Number UN1150
Beilstein 1420761
Merck Index 14,92
Solubility Information Miscible with ethanol,ethyl ether,acetone,benzene and chloroform. Immiscible with water.
Formula Weight 96.94
Percent Purity 98%
Chemical Name or Material trans-1,2-Dichloroethylene, stabilized with 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.