missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichloroethylene, 99+%, stabilized

Product Code. 10558892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10558892 250 g Glass bottle 250g
10481011 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10558892 Supplier Thermo Scientific Acros Supplier No. 406842500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 156-60-5 | C2H2Cl2 | 96.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-60-5
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.94
MDL Number MFCD00062942
InChI Key KFUSEUYYWQURPO-OWOJBTEDSA-N
Synonym trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
PubChem CID 638186
ChEBI CHEBI:29027
IUPAC Name (E)-1,2-dichloroethene
SMILES Cl\C=C\Cl

Specifications

Melting Point -50°C
Color Colorless
Density 1.2600g/mL
Boiling Point 48°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CHClCHCl
Refractive Index 1.4450 to 1.447
Quantity 250 g
Beilstein 01, 188
Merck Index 15, 87
Specific Gravity 1.26
Solubility Information Solubility in water: immiscible
Stabilizer 85 to 115ppm MEHQ
Formula Weight 96.94
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material trans-1, 2-Dichloroethylene, 99.7%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-1,2-Dichloroethylene, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.