missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Diaminocyclohexane, 98+%

Product Code. p-3736112 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10374742 25 g Plastic bottle 25g
10631853 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10374742 Supplier Thermo Scientific Supplier No. 406360250
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2615-25-0 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2615-25-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00035862,MFCD00075174,MFCD08276323
InChI Key VKIRRGRTJUUZHS-UHFFFAOYSA-N
Synonym trans-1,4-diaminocyclohexane, trans-cyclohexane-1,4-diamine, 1,4-diaminocyclohexane, trans-1,4-cyclohexanediamine, cis-cyclohexane-1,4-diamine, 1,4-cyclohexanediamine, cis-1,4-cyclohexanediamine, 1,4-cyclohexanediamine, trans, 1r,4r-cyclohexane-1,4-diamine, 1,4-cyclohexanediamine, cis
PubChem CID 18374
IUPAC Name cyclohexane-1,4-diamine
SMILES NC1CCC(N)CC1

Specifications

Melting Point 68.0°C to 71.0°C
Color White to Brown
Boiling Point 197.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H10(NH2)2
Quantity 25 g
Formula Weight 114.19
Percent Purity 99%
Physical Form Crystals and/or Chunks
Chemical Name or Material trans-1, 4-Diaminocyclohexane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-1,4-Diaminocyclohexane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.