Learn More
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane, 97%
Beskrivning
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.
Solubility
Slightly Soluble (4.4 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. Keep container tightly closed. Store away from oxidizing agent.
Specifikationer
Specifikationer
| Solubility Information | Slightly Soluble (4.4g/L) (25°C). |
| Quantity | 1 g |
| Formula Weight | 229.31 |
| Percent Purity | 97% |
| Physical Form | Solid |
| Chemical Name or Material | trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane |
RUO – Research Use Only
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.