missing translation for 'onlineSavingsMsg'
Learn More

trans-1-Hexen-1-ylboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15419377
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419377 1 g 1g
15429377 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419377 Supplier Thermo Scientific Alfa Aesar Supplier No. H52613.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42599-18-8 | Molecular Formula: C6H13BO2 | Molecular Weight: 127.978 g/mol

trans-1-Hexen-1-ylboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1-Hexen-1-ylboronic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42599-18-8
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.978
MDL Number MFCD01074553
InChI Key GWFKSQSXNUNYAC-AATRIKPKSA-N
Synonym e-hexen-1-ylboronic acid, e-hex-1-en-1-ylboronic acid, trans-1-hexen-1-ylboronic acid, boronic acid, b-1-hexen-1-yl, 1e-hex-1-en-1-ylboronic acid, boronic acid,b-1e-1-hexenyl, e-hex-1-enylboronic acid, e-1-hexenyldihydroxyborane, hex-1-en-1-ylboronic acid
PubChem CID 5326225
IUPAC Name [(E)-hex-1-enyl]boronic acid
SMILES B(C=CCCCC)(O)O

Specifications

Melting Point 62°C to 63°C
Quantity 1 g
Formula Weight 127.98
Percent Purity 98%
Chemical Name or Material trans-1-Hexen-1-ylboronic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.