missing translation for 'onlineSavingsMsg'
Learn More

trans-1-Octenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11315535
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315535 1 g 1g
11325535 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11315535 Supplier Thermo Scientific Alfa Aesar Supplier No. L19697.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83947-55-1
Molecular Formula C14H27BO2
Molecular Weight (g/mol) 238.178
MDL Number MFCD03453667
InChI Key KQTOSGTXAFJZSJ-VAWYXSNFSA-N
Synonym trans-4,4,5,5-tetramethyl-2-oct-1-enyl-1,3,2-dioxaborolane, 1-octenylboronic acid pinacol ester, 1-octen-1-ylboronic acid, pinacol ester, trans-1-octenylboronic acid pinacol ester, trans-4,4,5,5-tetramethyl-2-1-octenyl-1,3,2-dioxaborolane, e-1-octenylboronic acid pinacol ester, trans-1-octen-1-ylboronic acid pinacol ester, trans-1-oct-1-en-1-yl boronic acid, pinacol ester, trans-4,4,5,5-tetramethyl-2-oct-1-en-1-yl-1,3,2-dioxaborolane
PubChem CID 5706555
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-oct-1-enyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=CCCCCCC

Specifications

Refractive Index 1.446
Quantity 1 g
Beilstein 7291389
Formula Weight 238.18
Percent Purity 97%
Chemical Name or Material trans-1-Octenylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.