missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Aminocyclohexanol hydrochloride, 99%

Product Code. 10636602 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636602 5 g Glass bottle 5g
10289153 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10636602 Supplier Thermo Scientific Acros Supplier No. 206980050
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5456-63-3 | C6H13ClNO | 150.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5456-63-3
Molecular Formula C6H13ClNO
Molecular Weight (g/mol) 150.63
MDL Number MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963
InChI Key SEBSRZIHRZRNAH-KGZKBUQUSA-N
PubChem CID 12228412
SMILES [Cl].N[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 173.0°C to 177.0°C
Color Beige to Brown or White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H10OH·HCl
Quantity 5 g
Beilstein 13, III, 704
Synonym trans-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hydrochloride, 1r, 2r-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hcl, 1r,2r-trans-2-aminocyclohexanol hydrochloride, trans-2-hydroxycyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 2-amino-, hydrochloride, 1r,2r-rel, 1r,2r-2-aminocyclohexanolhydrochloride, 1r,2r-rel-2-aminocyclohexanol hydrochloride
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,ether,benzene
IUPAC Name (1R,2R)-2-aminocyclohexan-1-ol;hydrochloride
Formula Weight 151.64
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material trans-2-Aminocyclohexanol hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.