missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Dimethylamino)cyclopentanol, TRC

Product Code. 30385315
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385315 50 mg 50mg
30376586 500 mg -
30368860 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385315 Supplier Toronto Research Chemicals Supplier No. TRCD49574850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18760-79-7
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.2
IUPAC Name (1R,2R)-2-(dimethylamino)cyclopentan-1-ol
SMILES CN(C)[C@@H]1CCC[C@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H15NO/c1-8(2)6-4-3-5-7(6)9/h6-7,9H,3-5H2,1-2H3/t6-,7-/m1/s1
Formula Weight 129.1154
Chemical Name or Material trans-2-(Dimethylamino)cyclopentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.