missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Ethoxyethenyl-1-boronic acid pinacol ester, 95%

Product Code. 15458425
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458425 1 g 1g
15468425 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458425 Supplier Thermo Scientific Alfa Aesar Supplier No. H33367.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1201905-61-4 | C10H19BO3 | 198.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1201905-61-4
Molecular Formula C10H19BO3
Molecular Weight (g/mol) 198.07
MDL Number MFCD09702441,MFCD09998813
InChI Key MRAYNLYCQPAZJN-UHFFFAOYSA-N
Synonym e-2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-ethoxyvinylboronic acid pinacol ester, 2-e-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-boronic acid pinacol ester, 2-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-ylboronic acid pinacol ester, z-2-ethoxyethenyl boronic acid, pinacol ester, trans-2-ethoxyethenyl-1-boronic acid pinacol ester
PubChem CID 21973908
IUPAC Name 2-[(E)-2-ethoxyethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCOC=CB1OC(C)(C)C(C)(C)O1

Specifications

Boiling Point 50°C (0.3 mmHg)
Refractive Index 1.446
Quantity 1 g
Formula Weight 198.07
Percent Purity 98%
Chemical Name or Material trans-2-Ethoxyethenyl-1-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.