missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenyl acetate, 98%

Product Code. 11463068
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463068 250 g 250g
11443068 10 g 10g
11453068 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463068 Supplier Thermo Scientific Alfa Aesar Supplier No. A18786.30
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2497-18-9 | C8H14O2 | 142.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2497-18-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00009474
InChI Key HRHOWZHRCRZVCU-AATRIKPKSA-N
Synonym z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z
PubChem CID 2733294
IUPAC Name [(E)-hex-2-enyl] acetate
SMILES CCCC=CCOC(=O)C

Specifications

Density 0.9
Boiling Point 165°C to 166°C
Flash Point 58°C (136°F)
Refractive Index 1.427
Quantity 250 g
UN Number UN3272
Beilstein 1721851
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material trans-2-Hexenyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.