missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methyl-2-butenal, 97%

Product Code. 11387985
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11387985

Brand: Thermo Scientific Alfa Aesar L08113.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 497-03-0 | C5H8O | 84.118 g/mol

Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-03-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006977
InChI Key ACWQBUSCFPJUPN-HWKANZROSA-N
Synonym tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e
PubChem CID 5321950
IUPAC Name (E)-2-methylbut-2-enal
SMILES CC=C(C)C=O

Specifications

Density 0.871
Boiling Point 117°C to 119°C
Flash Point 18°C (64°F)
Odor Fruit-like
Refractive Index 1.448
Quantity 5 g
UN Number UN1989
Beilstein 1698207
Sensitivity Air sensitive
Solubility Information Soluble in water
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material trans-2-Methyl-2-butenal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.