missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenal, 96%

Product Code. 11392376
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11392376 5 g 5g
11302386 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11392376

Brand: Thermo Scientific Alfa Aesar L06108.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1576-87-0 | C5H8O | 84.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-87-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00009615
InChI Key DTCCTIQRPGSLPT-ONEGZZNKSA-N
Synonym trans-2-pentenal, 2-pentenal, e-2-pentenal, e-pent-2-enal, 3-ethylacrolein, pent-2-enal, 2-pentenal, e, 2-e-pentenal, 3-ethyl-2-propenal, e-pent-2-en-1-al
PubChem CID 5364752
IUPAC Name (E)-pent-2-enal
SMILES CCC=CC=O

Specifications

Density 0.86
Boiling Point 125°C
Flash Point 22°C (71°F)
Odor Fruit-like
Refractive Index 1.443
Quantity 5 g
UN Number UN1989
Beilstein 1719742
Sensitivity Air Sensitive
Formula Weight 84.12
Percent Purity 96%
Chemical Name or Material trans-2-Pentenal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.