missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenal, 97%

Product Code. 10063640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10063640 25 g Glass bottle 25g
10063630 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10063640 Supplier Thermo Scientific Acros Supplier No. 325420250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1576-87-0 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-87-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00009615
InChI Key DTCCTIQRPGSLPT-ONEGZZNKSA-N
Synonym trans-2-pentenal, 2-pentenal, e-2-pentenal, e-pent-2-enal, 3-ethylacrolein, pent-2-enal, 2-pentenal, e, 2-e-pentenal, 3-ethyl-2-propenal, e-pent-2-en-1-al
PubChem CID 5364752
IUPAC Name (E)-pent-2-enal
SMILES CCC=CC=O

Specifications

Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 115.0°C to 125.0°C
Flash Point 19°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CHOHC=CHC2H5
Refractive Index 1.4400 to 1.4470
Quantity 25 g
Beilstein 01, IV, 3459
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble
Formula Weight 84.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-2-Pentenal, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-2-Pentenal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.