missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Phenylcyclopropane-1-carboxylic acid, 97%

Product Code. 10627780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627780 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10627780 Supplier Thermo Scientific Acros Supplier No. 130460250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 939-90-2 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-90-2
MDL Number MFCD00001292
InChI Key AHDDRJBFJBDEPW-DTWKUNHWSA-N
Synonym trans-2-phenylcyclopropanecarboxylic acid, trans-2-phenyl-1-cyclopropanecarboxylic acid, 1r,2r-2-phenylcyclopropanecarboxylic acid, trans-2-phenylcyclopropane-1-carboxylic acid, 2-phcpcooh, 1r,2r-2-phenylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2-phenyl-, 1r,2r-rel, rac trans-2-phenylcyclopropanecarboxylic-d5 acid, pubchem22341, trans-2-phenylcyclopropancarboxylic acid
PubChem CID 237413
ChEBI CHEBI:79920
IUPAC Name (1R,2R)-2-phenylcyclopropane-1-carboxylic acid
SMILES C1C(C1C(=O)O)C2=CC=CC=C2

Specifications

Melting Point 86.0°C to 92.0°C
Color Beige or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C10H10O2
Linear Formula C6H5C3H4CO2H
Quantity 25 g
Beilstein 09,619
Molecular Weight (g/mol) 162.19
Formula Weight 162.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material trans-2-Phenylcyclopropane-1-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • trans-2-Phenylcyclopropane-1-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.