missing translation for 'onlineSavingsMsg'
Learn More

trans-3,4-Difluoro-4'-(4-n-pentylcyclohexyl)biphenyl, 97%, Thermo Scientific™

Product Code. 15402316
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15402316 1 g 1g
15412316 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15402316

Brand: Thermo Scientific Alfa Aesar H55835.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
trans-3,4-Difluoro-4′-(4-n-pentylcyclohexyl)biphenyl, is used to make PDLC film, smart film, switchable film, PDLC glass, smart glass, switchable glass, PDLC mixture. It also acts as a liquid crystal monomer.

Solubility
Sparingly Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134412-17-2
Molecular Formula C23H28F2
Molecular Weight (g/mol) 342.47
MDL Number MFCD09839000
InChI Key NQLHGECGZRJQLF-UHFFFAOYSA-N
Synonym 3,4-difluoro-4'-trans-4-pentylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-pentylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-pentylcyclohexyl phenyl benzene, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl, 1,1'-biphenyl,3,4-difluoro-4'-trans-4-pentylcyclohexyl, 3,4-difluoro-4'-1s,4r-4-pentylcyclohexyl-1,1'-biphenyl, acmc-20mvco, 3,4-difluoro-4/'-4-pentylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-pentylcyclohexyl benzene
PubChem CID 612307
IUPAC Name 1,2-difluoro-4-[4-(4-pentylcyclohexyl)phenyl]benzene
SMILES CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1

Specifications

Melting Point 56°C to 57°C
Quantity 1 g
Solubility Information Sparingly Soluble in water.
Formula Weight 342.47
Percent Purity 97%
Chemical Name or Material trans-3,4-Difluoro-4'-(4-n-pentylcyclohexyl)biphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.