missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Benzyloxycarbonylamino)cyclohexanemethanol, 97%, Thermo Scientific™

Product Code. 15447906
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15447906

Brand: Thermo Scientific Alfa Aesar H62842.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1202411-95-7
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.337
MDL Number MFCD09864989
InChI Key QYHAKPLUUUMEFH-ZIAGYGMSSA-N
Synonym benzyl trans-3-hydroxymethylcyclohexylcarbamate, benzyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
PubChem CID 59401466
IUPAC Name benzyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
SMILES C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)CO

Specifications

Quantity 1 g
Formula Weight 263.33
Percent Purity 97%
Chemical Name or Material trans-3-(Benzyloxycarbonylamino)cyclohexanemethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.