missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Boc-amino)cyclohexanemethanol, 97%

Product Code. 15449378
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15449378

Brand: Thermo Scientific Alfa Aesar H62934.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 920966-17-2 | C12H23NO3 | 229.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920966-17-2
Molecular Formula C12H23NO3
Molecular Weight (g/mol) 229.32
MDL Number MFCD03844607
InChI Key LBJSEPNOVVUVJA-NXEZZACHSA-N
Synonym tert-butyl trans-3-hydroxymethyl-cyclohexylcarbamate, tert-butyl trans-3-hydroxymethyl cyclohexylcarbamate, tert-butyl trans-3-hydroxymethylcyclohexyl carbamate, tert-butyl trans-3-hydroxymethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
PubChem CID 24720931
IUPAC Name tert-butyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)CO

Specifications

Quantity 250 mg
Formula Weight 229.31
Percent Purity 97%
Chemical Name or Material trans-3-(Boc-amino)cyclohexanemethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.