missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Chloroacrylic acid, 99%

Product Code. 11463920
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463920 250 mg 250mg
11432947 1 g 1g
11442947 5 g 5g
11452947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11463920 Supplier Thermo Scientific Alfa Aesar Supplier No. A15527.MD
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2345-61-1 | C3H3ClO2 | 106.505 g/mol

trans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.

Solubility
Soluble in water.

Notes
Store at 4°C. Store away from oxidizing agent, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2345-61-1
Molecular Formula C3H3ClO2
MDL Number MFCD00064237
InChI Key MHMUCYJKZUZMNJ-OWOJBTEDSA-N
Synonym trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546
PubChem CID 638124
IUPAC Name (E)-3-chloroprop-2-enoic acid

Specifications

Melting Point 81°C to 85°C
Quantity 250 mg
UN Number UN3261
Beilstein 1720279
Solubility Information Soluble in water.
SMILES C(=CCl)C(=O)O
Molecular Weight (g/mol) 106.505
Formula Weight 106.51
Percent Purity 99%
Chemical Name or Material trans-3-Chloroacrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.