missing translation for 'onlineSavingsMsg'
Learn More

trans-3-chloroacrylic acid, 99%

Product Code. 10774021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10774021 1 g Glass bottle 1g
10197793 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10774021 Supplier Thermo Scientific Acros Supplier No. 108550010
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2345-61-1 | C3H3ClO2 | 106.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2345-61-1
Molecular Formula C3H3ClO2
Molecular Weight (g/mol) 106.51
MDL Number MFCD00064237
InChI Key MHMUCYJKZUZMNJ-OWOJBTEDSA-N
Synonym trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546
PubChem CID 638124
IUPAC Name (E)-3-chloroprop-2-enoic acid
SMILES C(=CCl)C(=O)O

Specifications

Melting Point 82°C to 85°C
Color Beige-Gray to Gray
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClCH=CHCO2H
Quantity 1 g
Beilstein 02, 400
Formula Weight 106.51
Percent Purity 99%
Physical Form Powder or Flakes
Chemical Name or Material trans-3-chloroacrylic acid
Show More Show Less
compliance-icons
Product Identifier
  • trans-3-Chloroacrylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.